Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABI1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10059843PCP
Description
ABI1 Polyclonal antibody specifically detects ABI1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)Specifications
ABI1 | |
Polyclonal | |
Immunoprecipitation, Western Blot (Negative) | |
Abelson interactor 1, ABI-1, abl interactor 1, Abl-binding protein 4, AblBP4, abl-interactor 1, Abl-interactor protein 1 long, E3B1, eps8 binding protein, Eps8 SH3 domain-binding protein, Eps8-binding protein, interactor protein AblBP4, nap1 binding protein, Nap1-binding protein, Nap1BP, spectrin SH3 domain binding protein 1, Spectrin SH3 domain-binding protein 1, SSH3BP, SSH3BP1 | |
Maps to a region between residue 150 and 200 of human abl-interactor 1 using the numbering given in entry NP_005461.2 (GeneID 10006). | |
0.1 mL | |
Phospho Specific | |
10006 | |
Store at 4C in the dark. | |
IgG |
Immunoprecipitation | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction