Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570529
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570529 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570529 Supplier Invitrogen™ Supplier No. PA570529
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The nucleolar protein encoded by this gene interacts in a hormone-dependent manner with nuclear receptors, and represses their transcriptional activity. Alternative splice variants that encode different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABI1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ABI1
Gene Accession No. Q9QZM5
Gene Alias abelson interactor 1; Abi1; Abi-1; Abl interactor 1; abl-binding protein 4; AblBP4; abl-interactor 1; Abl-interactor protein 1 long; Ablphilin-1; E3b1; EL2; eps8 binding protein; eps8 binding protein (e3B1), alternatively spliced; eps8 SH3 domain-binding protein; Eps8-binding protein; HPP; HS3; interactor protein AblBP4; LOW QUALITY PROTEIN: abl interactor 1; NAP1; nap1 binding protein; Nap1-binding protein; Nap1BP; spectrin SH3 domain binding protein 1; spectrin SH3 domain-binding protein 1; SPH3; SPTA; SSH3BP; Ssh3bp1
Gene Symbols ABI1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ABI1 (aa 91-140).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79249
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.