Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABI1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571742
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1742 10 μL
50-157-1741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1742 Supplier Bethyl Laboratories, Inc Supplier No. IHC00240T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Chicken, Turkey, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Monke

ABI1 has been found to form a complex with EPS8 and SOS1, and is thought to be involved in the transduction of signals from Ras to Rac. In addition, this protein may play a role in the regulation of EGF-induced Erk pathway activation as well as cytoskeletal reorganization and EGFR signaling.

Specifications

Antigen ABI1
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ABI1
Gene Accession No. Q8IZP0
Gene Alias EL2, HPP, HS3, SPH3, SPTA
Gene Symbols ABI1
Host Species Rabbit
Immunogen Between 325 and 375
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10006
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.