Learn More
Abi1 Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$178.35 - $661.74
Specifications
| Antigen | Abi1 |
|---|---|
| Concentration | 1.07 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2087
![]()
|
Rockland Immunochemicals
600401GT5 |
100 μg |
Each for $661.74
|
|
|||||
50-199-2088
![]()
|
Rockland Immunochemicals
600401GT5S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Abi1 is directed against mouse Abi1. A BLAST analysis was used to suggest cross reactivity with this protein in human and rat based on 100% homology for the immunogen sequence. Reactivity against homologues from other sources is not known.
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The nucleolar protein encoded by this gene interacts in a hormone-dependent manner with nuclear receptors, and represses their transcriptional activity. Alternative splice variants that encode different isoforms have been found for this gene.Specifications
| Abi1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Q8CBW3 | |
| 11308 | |
| Primary | |
| Antibody |
| 1.07 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| Abelson interactor 1; ABI1; Abi-1; Abl interactor 1; Abl-binding protein 4; AblBP4; abl-interactor 1; Abl-interactor protein 1 long; Ablphilin-1; BOS_12878; e3B1; EL2; eps8 binding protein; eps8 binding protein (e3B1), alternatively spliced; Eps8 SH3 domain-binding protein; Eps8-binding protein; HPP; HS3; interactor protein AblBP4; LOW QUALITY PROTEIN: abl interactor 1; NAP1; nap1 binding protein; Nap1-binding protein; Nap1BP; spectrin SH3 domain binding protein 1; Spectrin SH3 domain-binding protein 1; SPH3; SPTA; SSH3BP; Ssh3bp1 | |
| ABI1 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| ABI1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.