Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595121
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595121 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595121 Supplier Invitrogen™ Supplier No. PA595121
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human 293T whole cell, human Hela whole cell, human K562 whole cell, human SH-SY5Y whole cell, rat brain tissue, rat PC-12 whole cell, mouse brain tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

ABI2 may act in regulation of cell growth and transformation by interacting with nonreceptor tyrosine kinases ABL1 and/or ABL2. ABI2 is a part of the WAVE complex that regulates lamellipodia formation. The WAVE complex regulates actin filament reorganization via its interaction with the Arp2/3 complex. It regulates ABL1/c-Abl-mediated phosphorylation of MENA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABI2
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene Abi2
Gene Accession No. P62484, Q9NYB9
Gene Alias 8430425M24Rik; Abelson interactor 2; Abi2; abi-2; ABI2B; abl binding protein 3; abl interactor 2; Abl-binding protein 3; AblBP3; abl-interacting protein 1 (SH3-containing protein); abl-interactor 2; abl-interactor protein 2b; AI839867; AIP-1; arg protein tyrosine kinase-binding protein; arg-binding protein 1; ArgBP1; ARGBPIA; argBPIB; C130078H13; SSH3BP2; thyroid hormone responsive protein
Gene Symbols Abi2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human ABI2 (158-173aa RFKVSTQNMKMGGLPR).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10152, 286928, 329165
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.