Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABI2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS5984R

Description
ABI2 may act in regulation of cell growth and transformation by interacting with nonreceptor tyrosine kinases ABL1 and/or ABL2. ABI2 is a part of the WAVE complex that regulates lamellipodia formation. The WAVE complex regulates actin filament reorganization via its interaction with the Arp2/3 complex. It regulates ABL1/c-Abl-mediated phosphorylation of MENA.Specifications
ABI2 | |
Polyclonal | |
Unconjugated | |
Abi2 | |
8430425M24Rik; Abelson interactor 2; ABI2; Abi-2; ABI2B; abl binding protein 3; Abl interactor 2; Abl-binding protein 3; AblBP3; abl-interacting protein 1 (SH3-containing protein); abl-interactor 2; abl-interactor protein 2b; AI839867; AIP-1; arg protein tyrosine kinase-binding protein; Arg-binding protein 1; ArgBP1; ARGBPIA; argBPIB; BOS_1973; C130078H13; SSH3BP2; thyroid hormone responsive protein | |
Rabbit | |
Protein A | |
RUO | |
10152 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9NYB9 | |
Abi2 | |
KLH conjugated synthetic peptide derived from human ABI2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction