Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABI3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553592
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553592 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553592 Supplier Invitrogen™ Supplier No. PA553592
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AYQVGNLAGH TLRMLDLQGA ALRQVEARVS TLGQMVNMHM EKVARREIGT LATVQRLPPG QKVIAPENLP PLTPYCRRPL NFGCLDDIGH GIKDLSTQLS RT Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.

Inhibits ectopic tumor cell metastasis of SRD cells. In vitro, reduces cell motility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABI3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABI3
Gene Accession No. Q9P2A4
Gene Alias 2210414K06Rik; ABI family member 3; ABI family, member 3; ABI gene family member 3; ABI gene family, member 3; Abi3; AI987680; NESH; NESH protein; new molecule including SH3; SSH3BP3
Gene Symbols ABI3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABI3. Recombinant protein control fragment (Product #RP-90723).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51225
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.