Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABI3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113400
Description
ABI3 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Inhibits ectopic tumor cell metastasis of SRD cells. In vitro, reduces cell motility.
Specifications
| ABI3 | |
| Polyclonal | |
| Unconjugated | |
| ABI3 | |
| 2210414K06Rik; ABI family member 3; ABI family, member 3; ABI gene family member 3; ABI gene family, member 3; Abi3; AI987680; NESH; NESH protein; new molecule including SH3; SSH3BP3 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 51225, 66610 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8BYZ1, Q9P2A4 | |
| ABI3 | |
| Recombinant Human ABI gene family member 3 protein (1-366AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction