Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABI3BP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542983
Description
Peptide sequence: PVSNTVAFST ESADPRVSEP VSAGRDAIWT ERPFNSDSYS ECKGKQYVKR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%; Zebrafish: 100%.
ABI3BP is a protein coding gene. Gene ontology (GO) annotation include collagen-containing extracellular matrix; extracellular region; extracellular space; interstitial matrix.
Specifications
| ABI3BP | |
| Polyclonal | |
| Unconjugated | |
| ABI3BP | |
| 5033411B22Rik; ABI family member 3 binding protein; ABI family, member 3 (NESH) binding protein; ABI gene family member 3-binding protein; ABI gene family, member 3 (NESH) binding protein; ABI3BP; AI506287; BG172926; D930038M13Rik; eratin; FLJ41743; FLJ41754; Nesh-binding protein; NESHBP; RGD1562717; Target of Nesh-SH3; Tarsh | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 25890 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q7Z7G0 | |
| ABI3BP | |
| Synthetic peptide directed towards the C-terminal of human ABI3BP (aa 914-963). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction