Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABI3BP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589914
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589914 Supplier Invitrogen™ Supplier No. PA589914
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QKLPKGKRPN LKVHINTTSD SILLKFLRPS PNVKLEGLLL GYGSNVSPNQ YFPLPAEGKF TEAIVDAEPK YLIVVRPAPP PSQKKSCSGK TRSRKPLQLV VGTLTPSSVF LSWGFLINPH HDWTLPSHCP NDRFYTIRYR EKDKEKKWIF QICPATETIV ENLKPNTVYE FGVKDNVEGG IWSKIFNHKT VVGSKKVNGK IQSTYDQDHT VPAYVPRKLI PITIIKQVIQ NVTHKDSAKS PEKAPLGGVI LV; Positive Samples: U-87MG, A-549, LO2, 293T, HeLa.

ABI3BP is a protein coding gene. Gene ontology (GO) annotation include collagen-containing extracellular matrix; extracellular region; extracellular space; interstitial matrix.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABI3BP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ABI3BP
Gene Accession No. Q7Z7G0
Gene Alias 5033411B22Rik; ABI family member 3 binding protein; ABI family, member 3 (NESH) binding protein; ABI gene family member 3-binding protein; ABI gene family, member 3 (NESH) binding protein; ABI3BP; AI506287; BG172926; D930038M13Rik; eratin; FLJ41743; FLJ41754; Nesh-binding protein; NESHBP; RGD1562717; Target of Nesh-SH3; Tarsh
Gene Symbols ABI3BP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 29-280 of human ABI3BP (NP_0562442).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25890
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.