Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ABI3BP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583706
Description
Immunogen sequence: VEGGIWSKIF NHKTVVGSKK VNGKIQSTYD QDHTVPAYVP RKLIPITIIK QVIQNVTHKD SAKSPEKAPL GGVILVHLII PGLNETTVKL PASLMFEISD AL.
ABI3BP is a protein coding gene. Gene ontology (GO) annotation include collagen-containing extracellular matrix; extracellular region; extracellular space; interstitial matrix.
Specifications
ABI3BP | |
Polyclonal | |
Unconjugated | |
ABI3BP | |
5033411B22Rik; ABI family member 3 binding protein; ABI family, member 3 (NESH) binding protein; ABI gene family member 3-binding protein; ABI gene family, member 3 (NESH) binding protein; ABI3BP; AI506287; BG172926; D930038M13Rik; eratin; FLJ41743; FLJ41754; Nesh-binding protein; NESHBP; RGD1562717; Target of Nesh-SH3; Tarsh | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25890 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z7G0 | |
ABI3BP | |
Recombinant protein corresponding to Human ABI3BP. Recombinant protein control fragment (Product #RP-100173). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction