Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abietic acid, tech. 75%

Catalog No. aal1377014
Click to view available options
Quantity:
25 g
100 g

CAS: 514-10-3 | C20H30O2 | 302.46 g/mol

Abietic acid is a useful precursor for synthesis of various natural products including diterpenes and steroids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Abietic acid is a useful precursor for synthesis of various natural products including diterpenes and steroids.

Solubility
Soluble in alcohols, acetone and ethers

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C. Store under inert gas. Sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 514-10-3
Molecular Formula C20H30O2
Molecular Weight (g/mol) 302.46
MDL Number MFCD03423567
InChI Key RSWGJHLUYNHPMX-ONCXSQPRSA-N
Synonym abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183
PubChem CID 10569
ChEBI CHEBI:28987
IUPAC Name (1R,4aR,4bR,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid
SMILES CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1

Specifications

Beilstein 2221451
Merck Index 14,7
Solubility Information Soluble in alcohols,acetone and ethers
Formula Weight 302.45
Percent Purity 75%
Grade Technical
Quantity 25 g
Odor Characteristic
Sensitivity Air sensitive
Chemical Name or Material Abietic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 208-178-3
RTECSNumber TP8580000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.