Learn More
Promega ABL1 Kinase Enzyme System, 1mg
Catalog No. PRV1902 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV1902 1 mg
1 options
Catalog No. PRV1902 Supplier Promega Supplier No. V1902
Add to Cart
Add to Cart
Request a Quote
Recombinant human ABL1 was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. ABL1 proto-oncogene encodes a cytoplasmic and nuclear protein tyrosine kinase implicated in processes of cell differentiation, cell division and others.
  • Easily Screen and Profile ABL1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C