Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABL2 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP118875B
Description
ABL2 Polyclonal antibody specifically detects ABL2 in Human, Mouse samples. It is validated for Western BlotSpecifications
ABL2 | |
Polyclonal | |
Western Blot | |
Abelson murine leukemia viral (v-abl) oncogene homolog 2, Abelson murine leukemia viral oncogene homolog 2, Abelson-related gene protein, Abelson-relatedgene), arg tyrosine kinase, EC 2.7.10, EC 2.7.10.2, FLJ22224, FLJ31718, FLJ41441, tyrosine-protein kinase ABL2, Tyrosine-protein kinase ARG, v-abl Abelson murine leukemia viral oncogene homolog 2 | |
The immunogen recognized by this antibody maps to a region between residue 750 and 800 of human Abelson-related gene protein using the numbering given in entry NP_009298.1 (GeneID 27). | |
0.1 mL | |
Protein Kinase | |
27 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction