Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Abl2 Monoclonal Antibody (1H1B11)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515330
Description
MA5-15330 targets ABL2 in indirect ELISA, ICC and WB applications and shows reactivity with Human samples. The MA5-15330 immunogen is purified recombinant fragment of ABL2 expressed in E. Coli.
ABL2 (Arg) is an Abl-related non-receptor protein tyrosine kinase that links cell surface receptors to cell proliferation and/or cytoskeletal reorganization. Abl2 encodes a member of the Abelson family of nonreceptor tyrosine protein kinase. The protein is highly similar to the ABL1 protein, including the tyrosine kinase, SH2 and SH3 domains, and has a role in cytoskeletal rearrangements by its C-terminal F-actin- and microtubule-binding sequences. Abl2 is expressed in both normal and tumor cells, and is involved in translocation with the ETV6 gene in leukemia. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene.
Specifications
Abl2 | |
Monoclonal | |
Conc. Not Determined | |
Ascites with 0.03% sodium azide | |
P42684 | |
ABL2 | |
Purified recombinant fragment of ABL2 expressed in E. Coli. | |
RUO | |
27 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1H1B11 | |
Unconjugated | |
ABL2 | |
AA536808; Abelson murine leukemia viral (v-abl) oncogene homolog 2; abelson murine leukemia viral oncogene homolog 2; Abelson tyrosine-protein kinase 2; Abelson-related gene protein; ABL proto-oncogene 2, non-receptor tyrosine kinase; Abl1-related gene product; abl2; ABLL; ARG; arg tyrosine kinase; c-abl oncogene 2, non-receptor tyrosine kinase; FLJ22224; FLJ31718; FLJ41441; tyrosine kinase ARG; tyrosine-protein kinase ABL2; Tyrosine-protein kinase ARG; v-abl Abelson murine leukemia viral oncogene 2; v-abl Abelson murine leukemia viral oncogene 2 (arg, Abelson-related gene); v-abl Abelson murine leukemia viral oncogene homolog 2; v-abl Abelson murine leukemia viral oncogene homolog 2 (arg, Abelson-related gene) | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction