Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABLIM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112346
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112346 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112346 Supplier Invitrogen™ Supplier No. PA5112346
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABLIM1 Polyclonal Antibody for Western Blot

This gene encodes a cytoskeletal LIM protein that binds to actin filaments via a domain that is homologous to erythrocyte dematin. LIM domains, found in over 60 proteins, play key roles in the regulation of developmental pathways. LIM domains also function as protein-binding interfaces, mediating specific protein-protein interactions. The protein encoded by this gene could mediate such interactions between actin filaments and cytoplasmic targets. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABLIM1
Applications Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ABLIM1
Gene Accession No. O14639, Q8K4G5
Gene Alias 2210411C18Rik; 2610209L21Rik; 4833406P10Rik; 9330196J19Rik; ABLIM; ABLIM1; abLIM-1; actin binding LIM protein 1; actin-binding double zinc finger protein; actin-binding double-zinc-finger protein; actin-binding LIM protein 1; Actin-binding LIM protein family member 1; AV079770; AW060987; AW215784; KIAA0059; LIMAB1; Limatin; mKIAA0059; RGD1565768
Gene Symbols ABLIM1
Host Species Rabbit
Immunogen Recombinant Human Actin-binding LIM protein 1 protein (174-319AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226251, 3983
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.