Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABLIM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557405
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557405 Supplier Invitrogen™ Supplier No. PA557405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGEDGSLDQD NRKKSSWLML KGDADTRTNS PDLDTQSLSH SSGTDRDPLQ RMAGDSFHSR FPYSKSDPLP GHGKNGLDQR NANLAPCGAD PDAS Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 78%.

ABLIM2 (Actin Binding LIM Protein Family Member 2) is a Protein Coding gene. Among its related pathways areDevelopmental BiologyandAxon guidance. Gene Ontology (GO) annotations related to this gene includeactin binding. An important paralog of this gene isABLIM1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABLIM2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ablim2
Gene Accession No. Q6H8Q1
Gene Alias Ablim2; abLIM-2; actin binding LIM protein 2; actin binding LIM protein family member 2; actin binding LIM protein family, member 2; actin-binding LIM protein 2; Actin-binding LIM protein family member 2; AI606905; C230091L11; KIAA1808; testis tissue sperm-binding protein Li 35a
Gene Symbols Ablim2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABLIM2. Recombinant protein control fragment (Product #RP-96714).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84448
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.