Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ABLIM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116761
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116761 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116761 Supplier Invitrogen™ Supplier No. PA5116761
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse lung, Rat brain Immunogen sequence: TSEDISQTSK YSPIYSPDPY YASESEYWTY HGSPKVPRAR RFSSGGEEDD FDRSMHKLQS GIGRLILKEE MKARSSSYAD PWTPPRSSTS SREALHTAGY EMSLNGSPRS HYLADSDPLI SKSASLPAYR RNGLHRTPSA DLFHYDSMNA VNWGMREYKI YPYELLLVTT R.

The LIM domain is a double zinc finger structure that promotes protein-protein interactions. LIM domain proteins, such as ABLIM3, play roles in embryonic development, cell lineage determination, and cancer (Krupp et al., 2006 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABLIM3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ablim3
Gene Accession No. Q69ZX8
Gene Alias ABLIM3; abLIM-3; actin binding LIM protein family member 3; actin binding LIM protein family, member 3; actin-binding LIM protein 3; Actin-binding LIM protein family member 3; AW990719; D930036B08Rik; HMFN1661; KIAA0843; RGD1565118
Gene Symbols Ablim3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 350-520 of human ABLIM3 (NP_0012879571).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307395, 319713
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.