Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abn-CBD, Tocris Bioscience™
SDP

Catalog No. 129710 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-971-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-971-0 Supplier R&D Systems Supplier No. 1297/10

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective GPR55 agonist; neurobehaviorally inactive

Neurobehaviorally inactive cannabinoid that acts as a selective agonist for GPR55 (EC50 values are 2.5, >30 and >30 μM at GPR55, CB1 and CB2 receptors respectively). Increases phosphorylation of protein kinases in, and migration of, human umbilical vein endothelial cells.

Chemical Identifiers

CAS 22972-55-0
Molecular Formula C21H30O2
Molecular Weight (g/mol) 314.469
InChI Key YWEZXUNAYVCODW-RBUKOAKNSA-N
Synonym abnormal cannabidiol, abn-cbd, 4-1r,6r-3-methyl-6-prop-1-en-2-yl cyclohex-2-en-1-yl-5-pentylbenzene-1,3-diol, e---4-p-mentha-1,8-dien-3-yl-5-pentylresorcinol, resorcinol, 4-p-mentha-1,8-dien-3-yl-5-pentyl-,--e, abnormal-cannabidiol, d0s3um, 4-1r,6r-3-methyl-6-1-methylethenyl-2-cyclohexen-1-yl-5-pentyl-1,3-benzenediol, 4-1r,6r-3-methyl-6-prop-1-en-2-yl-1-cyclohex-2-enyl-5-pentylbenzene-1,3-diol, 4-1r,6r-3-methyl-6-prop-1-en-2-ylcyclohex-2-en-1-yl-5-pentylbenzene-1,3-diol
PubChem CID 89949
IUPAC Name 4-[(1R,6R)-3-methyl-6-prop-1-en-2-ylcyclohex-2-en-1-yl]-5-pentylbenzene-1,3-diol
SMILES CCCCCC1=CC(=CC(=C1C2C=C(CCC2C(=C)C)C)O)O

Specifications

Quantity 10 mg
Formula Weight 314.47
Percent Purity >97%
Chemical Name or Material Abn-CBD
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.