Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ A2m (Rat) ELISA Kit

Catalog No. p-4392775 Shop All Abnova Corporation Products
Click to view available options
Sample Type:
CCS, Milk, Plasma, Saliva, Serum, Urine
Plasma, Serum

Enzyme-linked immunoassay for the quantitative determination of A2m in Rat biological samples

Specifications

Calibration Range 6.25 to 400ng/mL
Conjugate HRP
Cross Reactivity Rat
Description A2m (Rat) ELISA Kit is a highly sensitive two-site enzyme-linked immunoassay for the quantitative determination of A2m in rat biological samples.
Detection Method Colorimetric
Includes 50mL Diluent concentrate, 50mL wash solution concentrate, 150μL Enzyme Antibody Conjugate concentrate, 12mL TMB Substrate solution, 12mL stop solution, 96-well microtiter plate, 1 vial calibrator (lyophilized)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type A2m (Rat) ELISA Kit
No. of Assays 96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.