Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Albumin (Human) ELISA Kit

Catalog No. p-4392008
Click to view available options
Sample Type:
CCS, CSF, Milk, Saliva, Urine
Plasma, Serum

Includes

  • Human Albumin Microplate: A 96-well polystyrene microplate (12 strips of 8 wells) coated with a polyclonal antibody against human albumin - 1
  • Sealing Tapes: Pre-cut, pressure-sensitive sealing tapes, which can be cut to fit the format of the individual assay - 3
  • Human Albumin Standard: Human albumin in a buffered protein base ( lyophilized) 800ng
  • Biotinylated Human Albumin Antibody (50x): A 50-fold concentrated biotinylated polyclonal antibody against human albumin 140μL
  • MIX Diluent Concentrate (10x): A 10-fold concentrated buffered protein base 30mL
  • Wash Buffer Concentrate (20x): A 20-fold concentrated buffered surfactant 30mL, 2 bottles
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate 80μL
  • Stop Solution: A 0.5N hydroxychloric acid to stop the chromogen substrate reaction 12mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Human albumin

Specifications

Conjugate Biotin
Cross Reactivity Human
Description Albumin (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human albumin.
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 800ng Human Albumin standard, 1 vial Biotinylated Human Albumin Antibody, 30mL mix diluent concentrate (10X), 30mL x 2 Wash Buffer 20X, 80 Streptavidin-Peroxidase Conjugate (SP Conjugate), 8mL chromogen Substrate, 12mL stop solution
Quantity 1 Kit
Sample Type CCS, CSF, Milk, Saliva, Urine
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type Albumin (Human) ELISA kit
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.