Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ C3 (Human) ELISA Kit
Shop All Abnova Corporation Products
Click to view available options
Sample Type:
CCS, CSF, Milk, Saliva, Urine
Plasma, Serum
Description
Includes
- Human Complement C3 Microplate: A 96-well polystyrene microplate (12 strips of 8 wells) coated with a polyclonal antibody against human complement C3
- Sealing Tapes: Each kit contains 3 pre-cut, pressure-sensitive sealing tapes that can be cut to fit the format of the individual assay
- Human Complement C3 Standard: Human Complement C3 in a buffered protein base (60μg, lyophilized)
- Biotinylated Complement C3: 1 vial, lyophilized
- EIA Diluent Concentrate (10x): A 10-fold concentrated buffered protein base (30mL)
- Wash Buffer Concentrate (20x): A 20-fold concentrated buffered surfactant (30mL)
- Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate (μL)
- Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine (8mL)
- 9. Stop Solution: A 0.5 N hydrochloric acid to stop the chromogen substrate reaction (12mL)
Specifications
Specifications
Calibration Range | 0.625 to 40ng/mL |
Cross Reactivity | Human |
Description | C3 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human complement C3 in urine, milk, saliva, and cell culture supernatants. |
Detection Method | Colorimetric |
Quantity | 1 Kit |
Sample Type | CCS, CSF, Milk, Saliva, Urine |
Sample Volume | 50 μL |
Content And Storage | Store SP Conjugate and Biotinylated Antibody at -20°C. Store Standard at 2-8°C before reconstituting with Diluent and at -20°C after reconstituting with Diluent. Store all other components at 4°C. |
For Use With (Application) | Quantification |
Product Type | C3 (Human) ELISA kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction