Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CCL4 (Rat) ELISA Kit

Catalog No. 89101097 Shop All Abnova Corporation Products
Click to view available options
Quantity:
1 Kit

Catalog No. 89-101-097

Supplier: Abnova™ KA3685

Only null left
Add to Cart
Add to Cart

Includes

  • 96-well plate precoated with anti-rat CCL4 antibody 96 (8 x 12) wells
  • Lyophilized recombinant rat CCL4 standard, 10ng x 2
  • Biotinylated anti-rat CCL4 antibody (dilution 1:100), 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100), 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

Use to measure quantitatively rat Ccl4. Sandwich enzyme immunoassay.

Ccl4 (Rat) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Assay Sensitivity <10 pg/mL
Calibration Range 15.6 to 1000pg/mL
Content And Storage Store the kit at -20°C
Description Ccl4 (Rat) is a sandwich enzyme immunoassay for the quantitative measurement of rat Ccl4.
Conjugate Biotin
Cross Reactivity Rat
Detection Method Colorimetric
Product Type Ccl4 (Rat) ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Specificity Natural and recombinant rat CCL4
Sufficient For 96 Assays
For Use With (Application) Quantification
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Detection Limit <10pg/mL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.