Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CCL8 (Human) ELISA Kit

Catalog No. p-4392446
Click to view available options
Sample Type:
CCS, Plasma, Serum, Tissue Sample, Urine
CCS, Plasma, Serum, Urine

In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human CCL8

Specifications

Calibration Range 30 to 1000pg/mL
Conjugate HRP
Cross Reactivity Human
Description CCL8 (Human) ELISA Kit is intended for the quantitative measurement of human CCL8.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes 96-well plate pre-coated with anti-human MCP-2antibody, Standard (0.5mL), Standard diluent buffer (1.5mL), 30X wash buffer (20mL), Sample diluent buffer (6mL), HRP conjugated anti-human MCP-2 antibody (6mL), Stop solution (6mL), TMB Substrate A(6mL), TMB Substrate B (6mL), Plate sealer, Hermetic bag
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma, Serum, Tissue Sample, Urine
Sample Volume 10 μL
Content And Storage Store at 4°C for 6 months.
For Use With (Application) Quantification
Product Type CCL8 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.