Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cdh1 (Mouse) ELISA Kit

Catalog No. p-4392064
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Body Fluid, CCS, Serum, Tissue Lysates

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Cdh1

Specifications

Calibration Range 156 to 10000pg/mL
Conjugate Biotin
Cross Reactivity Human, Mouse
Description Mouse Cdh1 ELISA kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Cdh1.
Detection Limit 15pg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 10ng x 2 vials lyophilized recombinant Mouse Cdh1 standard, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Mouse Cdh1 Antibody,, 12mL Antibody Diluent Buffer,130μL Avidin Biotin Peroxidase Complex,12mL ABC diluent buffer, 10mL TMB color developing agent, 10mL TMB stop solution
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, CCS, Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type CDH1 (Mouse) ELISA kit
No. of Assays 96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.