Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cdh2 (Rat) ELISA Kit

Catalog No. 89139271
Click to view available options
Quantity:
1 Kit

Sandwich enzyme immunoassay for the quantitative measurement of rat Cdh2 in cell culture supernates, plasma (heparin, EDTA) and serum samples

  • Sample Type: Cell culture supernates, plasma (heparin, EDTA) and serum
  • Sample Volume: 100μL
  • Conjugate: Biotin
  • Reactivity: Rat
  • Detection Method: Colorimetric
  • Calibration Range: 0.78 to 50 ng/mL
  • Limit of Detection: <20 pg/mL

  • Applications: Quantification
  • Storage: Store the kit at -20°C.

Specifications

Assay Sensitivity <20 pg/mL
Calibration Range 0.78 to 50 ng/mL
Description Cdh2 (Rat) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of rat Cdh2.
Detectable Analytes Cdh2
Assay Sandwich ELISA
Conjugate Biotin
Cross Reactivity No detectable cross-reactivity with other relevant proteins
Detection Method Colorimetric
Immunoassay Kit Format 96-well Plate
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Specificity Natural and recombinant rat Cadherin-2
Sufficient For 96 Assays
For Use With (Application) Quantification
Gene ID (Entrez) 83501
Host Species Rat
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Storage Requirements Store the kit at -20°C
Show More Show Less

For research use only (RUO).

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.