Learn More
Specifications
Specifications
| Calibration Range | Colorimetric: 4 to 400μM, Fluorometric: 0.5 to 40μM |
| Content And Storage | Store the kit at -20°C. |
| Description | Citrate Assay Kit is a quantitative colorimetric/fluorimetric determination of citrate at OD570, FL530/585 nm. |
| For Use With (Application) | Functional Study |
| Quantity | 1 Kit |
| Sample Type | Culture Medium, Plasma, Serum, Tissue, Urine |
| Sample Volume | 20 μL |
| Type | Cellular Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.