Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cortisol (Sheep) ELISA Kit

Catalog No. p-4392168 Shop All Abnova Corporation Products
Click to view available options
Sample Type:
Plasma, Serum, Urine

Immunoassay for the quantitative determination of cortisol in sheep serum, plasma, urine

Specifications

Calibration Range 0 to 200ng/mL
Conjugate HRP
Cross Reactivity Sheep
Description Cortisol (Sheep) ELISA Kit is an immunoassay for the quantitative determination of cortisol in sheep serum, plasma, urine.
Detection Limit 1.0ng/mL
Detection Method Colorimetric
Includes 96 wells-Microtiter wells coated with cortisol specific antibody, HRPEnzyme Conjugate (12mL), Cortisol Standards 1 sets (0.5mL/vial), TMB Color reagent (12mL), Stop solution (6mL), Wash buffer 20X (25mL), Standard diluent (20mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum, Urine
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Cortisol (Sheep) ELISA kit
No. of Assays 96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.