Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CytoChipNano Combo Pack

Catalog No. 89139310 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-310 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-310 Supplier Abnova™ Supplier No. KA4440
Add to Cart
Add to Cart
Request a Quote

Includes

Each combo includes: CytoChipNano, SCx™ Spiral Chamber and Plugs (one Clean Chip is included for both Prime and Clean purposes, one Dummy Chip is included for Tension Alignment purpose if needed)

For use with CytoQuest CR.

  • For seperation of CRCs on CytoQuest™ CR
  • Contains 20 combos; Each combo includes: CytoChipNano, SCx™ Spiral Chamber and Plugs
Applications:
Immunofluorescence

Recommended Storage

Store CytoChipNano at 4°C and SCx Spiral Chamber at room temperature.

Specifications

Content And Storage Store CytoChipNano at 4°C and SCx Spiral Chamber at room temperature.
Includes Each combo includes: CytoChipNano, SCx™ Spiral Chamber and Plugs (one Clean Chip is included for both Prime and Clean purposes, one Dummy Chip is included for Tension Alignment purpose if needed)
Quantity 1 Kit
For Use With (Equipment) CytoQuest CR

For research use only (RUO).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.