Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ ENO2 (Human) ELISA Kit

Catalog No. p-4392629
Click to view available options
Sample Type:
Serum

Enzyme Immunoassay for the quantitative measurement of ENO2 in Serum

Specifications

Conjugate HRP
Cross Reactivity Human
Description ENO2 (Human) ELISA Kit is an enzyme immunoassay for the quantitative measurement of ENO2 in serum.
Detection Limit 0.19ng/mL
Detection Method Colorimetric
Includes Fourteen 0.75mL h-NSE standard and control vial, 50mL incubation Buffer, 0.6mL Conjugate, 96-Anti-Monoclonal h-NSE adsorbed on microplate well, 12mL TMB-Substrate, 12mL stop solution, 20mL Wash Buffer
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 25 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ENO2 (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.