Learn More
Description
Includes
- Adhesive foil ready for use, 4 x 2
- Wash buffer concentrate, Concentrate. Dilute content with dist. water to a final volume of 1000mL, 20mL x 2
- Enzyme conjugate ready for use, anti-rabbit IgG conjugated with peroxidase, 12mL x 2
- Substrate ready for use, containing a solution of tetramethylbenzidine (TMB), 12mL x 2
- Stop solution ready for use, containing 0.25M H2SO4, 12mL x 2
- Adrenaline - Metanephrine microtiter strips, break apart, pre-coated, blue coloured, 96 wells 12 strips, 8 wells each
- Noradrenaline - Normetanephrine microtiter strips, break apart, pre-coated, yellow coloured, 96 wells 12 strips, 8 wells each
- Adrenaline antiserum from rabbit, ready for use, blue coloured, blue screw cap, 6mL
- Noradrenaline antiserum from rabbit, ready for use, yellow coloured, yellow screw cap, 6mL
- Adjustment buffer ready for use, 4mL
- Standard A ready for use, 4mL
Specifications
Specifications
| Calibration Range | Epinephrine: 1.3 to 81.4ng/mL, Norepinephrine: 4.9 to 339ng/mL |
| Description | Epinephrine/Norepinephrine ELISA Kit is an enzyme immunoassay for the quantitative determination of Epinephrine and Norepinephrine in plasma and urine. This product contains two assay plates, for Epinephrine and Normetanephrine separately. |
| Detection Method | Colorimetric |
| Includes | Adhesive foil 4 x 2, 2 x 20mL Wash Buffer concentrate, 2 12mL Enzyme Conjugate, 2 12mL Substrate, 2 12mL stop solution, 96-well Adrenaline, 96-well Noradrenaline, 6mL Adrenaline antiserum from Rabbit, 6mL Noradrenaline antiserum from Rabbit, 4mL Adjustment Buffer, 4mL standard A, 4mL standard B, 4mL, standard C, 4mL standard D, 4mL standard E, 4mL standard F, 20mL Acylation Buffer, 3mL Acylation reagent, 6mL Assay Buffer, 4mL coenzyme, four 1mL Enzyme lyophilized, 6mL Extraction Buffer, 2 48-well extraction plate coated with boronate affinity gel, 20mL Hydrochloric acid, 4mL control 1, 4mL control 2 |
| Quantity | 1 Kit |
| Sample Type | Plasma, Urine |
| Sample Volume | 10 μL (Urine), 300 μL (Plasma) |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Quantification |
| Product Type | Epinephrine/Norepinephrine ELISA kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.