Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Fibrinogen (Rat) ELISA Kit

Catalog No. p-4392212
Click to view available options
Sample Type:
CCS, Urine
Plasma

Sandwich Enzyme Immunoassay for the quantitative measurement of Rat Fibrinogen in Plasma

Specifications

Calibration Range 500 to 10000ng/mL
Conjugate Biotin
Cross Reactivity Rat
Description Fibrinogen (Rat) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of rat fibrinogen in plasma.
Detection Limit 300ng/mL
Detection Method Colorimetric
Includes 96 Antibody coated microwell, 240μg Rat FBG standard, Biotinylated rat FBG vial, 30mL MIX diluent (10X), 30mL Wash Buffer (20X), 80μL Streptavidin Peroxidase Conjugate, 8mL Chromogen Substrate, 12mL stop solution
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma
Sample Volume 25 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type Fibrinogen (Rat) ELISA kit
No. of Assays 96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.