Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ FLT1 (Human) ELISA Kit

Catalog No. p-4392027
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
CCS, Plasma (EDTA), Serum

Sandwich Enzyme Immunoassay for the quantitative measurement of Human FLT1

Specifications

Calibration Range 31.2 to 2000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description sFLT1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human soluble FLT1.
Detection Method Colorimetric
Includes 96 wells Antibody coated microwell, 10ng lyophilized recombinant Mouse Fn1 standard vials, 30mL sample diluent Buffer, 130μL Biotinylated Anti-Mouse Fn1 Antibody, 12mL Antibody Diluent Buffer, 130μL Avidin Biotin Peroxidase c
Quantity 1 Kit
Sample Type CCS, Plasma (EDTA), Serum
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type FLT1 (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.