Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Fn1 (Mouse) ELISA Kit
Shop All Abnova Corporation Products
Click to view available options
Sample Type:
Biological Fluid, CCS, Plasma, Serum, Tissue Lysates
CCS, Plasma, Serum, Urine
Description
- Sample Type: Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
- Sample Volume: 100μL
Quantification
Specifications
Specifications
Calibration Range | 3.13 to 200ng/mL |
Cross Reactivity | Mouse |
Description | Fn1 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Fn1. |
Detection Method | Colorimetric |
Quantity | 1 Kit |
Sample Type | CCS, Plasma, Serum, Urine |
Sample Volume | 50 μL |
Content And Storage | Store components of the kit at 4°C or -20°C as described in the protocol. |
For Use With (Application) | Quantification |
Product Type | Fn1 (Mouse) ELISA kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction