Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ GDF15 (Human) ELISA Kit

Catalog No. p-4392793
Click to view available options
Quantity:
1 Kit
96 Assays
Sample Type:
CCS, Plasma (EDTA, Heparin), Serum
CCS, Plasma, Serum, Urine

In-vitro ELISA for the quantitative measurement of Human GDF15

Specifications

Calibration Range 31.2 to 2000pg/mL
Cross Reactivity Human
Description GDF15 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human GDF15.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type CCS, Plasma (EDTA, Heparin), Serum
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type GDF15 (Human) ELISA kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.