Learn More
Abnova™ Global Pan-Methyl Histone H4K20 Quantification Kit (Colorimetric)
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Calibration Range | 20 to 2000ng/well |
| Content And Storage | Store the standard at -20°C. Store all other components at 4°C away from light. |
| Description | Global Pan-Methyl Histone H4K20 Quantification Kit (Colorimetric) is used for measuring global pan-methylation of histone H4K20. |
| Detection Method | Colorimetric |
| For Use With (Application) | Quantification |
| Includes | 20mL C1 (wash buffer 10X), 12mL C2 (antibody buffer), 10μL C3 (detection antibody, 1mg/mL), 10mL C4 (color developer), 6mL C5 (stop solution), 20μL standard control (100μg/mL), 10μL s |
| Quantity | 1 Kit |
| Sample Type | Histone Extract |
| Sample Volume | 50 μL |
| Type | Quantification Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.