Learn More
Specifications
Specifications
| Calibration Range | 0.01 to 3μM |
| Content And Storage | Store the kit at -20°C. |
| Description | GSH/GSSG Assay Kit is a quantitative colorimetric determination of GSH/GSSG at 412 nm. |
| Detection Method | Colorimetric |
| For Use With (Application) | Functional Study |
| Quantity | 1 Kit |
| Sample Type | Cell Extract, Plasma, Serum, Tissue, Whole Blood, Urine |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.