Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Hp (Rat) ELISA Kit

Catalog No. p-4392210
Click to view available options
Sample Type:
CCS, Urine
Plasma, Serum

Sandwich Enzyme Immunoassay for the quantitative measurement of Rat Haptoglobulin in Plasma and Serum

Specifications

Calibration Range 1000 to 15000ng/mL
Conjugate Biotin
Cross Reactivity Rat
Description Haptoglobulin (Rat) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of rat haptoglobulin in plasma and serum.
Detection Limit 20ng/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 60μg Rat Haptoglobulin standard, 1 vial Biotinylated Haptoglobulin, 30mL MIX diluent concentrate (10X), 30mL Wash Buffer 20X, 80μL Streptavidin Peroxidase Conjugate (SP Conjugate), 8mL
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 25 μL
Content And Storage Store the kit at 4°C or -20°C.
For Use With (Application) Quantification
Product Type Hp (Rat) ELISA Kit
No. of Assays 96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.