Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Hydrogen Peroxide Colorimetric Detection Kit
Click to view available options
Quantity:
1 Kit
Description
Includes
- Half-Area microtiter plate, ready to use, 1 each
- Hydrogen Peroxide Standard, (A solution of Hydrogen Peroxide at 100,000ng/mL in water with preservatives), 0.5mL
- Hydrogen Peroxide Color Reagent, (A solution of colorimetric substrate in dilute acid), 11mL
- Plate Sealer, 2 each
- Measurement helps to determine how oxidative stress modulates varied intracellular pathways
- Complete kit for the determination of hydrogen peroxide in biological fluids and tissue culture media
- Designed to measure low concentrations of hydrogen peroxide in biological matrices
- All components of this kit, except the hydrogen peroxide color reagent, are stable at 4°C until the expiration date; color reagent must be stored at -20°C
Specifications
Specifications
Description | Hydrogen Peroxide Colorimetric Detection Kit is a higher throughput assay for the quantitative measurement of hydrogen peroxide in culture supernatants, serum, plasma, and urine from any species. |
Detection Method | Colorimetric |
Product Type | Hydrogen Peroxide Colorimetric Detection Kit |
Sample Type | Plasma, Serum, Tissue Culture Media, Urine |
For Use With (Application) | Quantification |
Kit Contents | Hydrogen peroxide color reagent 11 mL, Hydrogen peroxide standard 0.5 mL, Microwell 96-well |
Quantity | 1 Kit |
Sample Volume | 50 μL |
Storage Requirements | Store color reagent at -20°C. Store all other components at 4°C. Avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction