Learn More
Abnova™ Hydroxymethylated DNA Quantification Kit (Colorimetric)
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Calibration Range | 50 to 200ng |
| Content And Storage | Store HC3, HC4, HC5, HC7 and HC8 at -20°C away from light. Store HC1, HC6, HC9 and 8-Well Assay Strips at 4°C away from light. Store all other components at room temperature away from light. |
| Description | Hydroxymethylated DNA Quantification Kit (Colorimetric) is used for measuring hydroxymethylated DNA. |
| Detection Method | Colorimetric |
| For Use With (Application) | Quantification |
| Includes | 14mL HC1 (wash buffer 10X), 5mL HC2 (binding solution), 10μL HC3 (negative control I, 20μg/mL), 10μL HC4 (negative control II, 20μg/mL), 10μL HC5 (positive control, |
| Quantity | 1 Kit |
| Sample Type | DNA Extract |
| Sample Volume | 1 to 8 μL |
| Type | Quantification Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.