Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IgA (Human) ELISA Kit

Catalog No. p-4747528
Click to view available options
Sample Type:
Cell Culture Samples, CSF, Milk, Plasma, Saliva, Serum, Urine
Serum, Plasma
The IgA (Human) ELISA Kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring IgA in serum or plasma of Human’s

Specifications

Calibration Range 12.5 to 800ng/mL
Conjugate HRP
Cross Reactivity Human
Description IgA (Human) ELISA Kit is a two-site enzyme linked immunoassay for the measurement of human IgA.
Detection Method Colorimetric
Includes Diluent concentrate (50mL), Wash solution concentrate (50mL), Enzyme-Antibody Conjugate Concentrate (150μL), TMB substrate solution (12mL), Stop solution (12mL), Microtiter plate (96 well), Calibrator (Lyophilized): 1 vial
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum, Plasma
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.