Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Il10 (Mouse) ELISA Kit

Catalog No. p-4392077 Shop All Abnova Corporation Products
Click to view available options
Sample Type:
Body Fluid, CCS, Serum, Tissue Lysates
CCS, Plasma, Serum, Tissue

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse IL-10

Specifications

Calibration Range 31 to 2000pg/mL
Conjugate HRP
Cross Reactivity Mouse
Description Il10 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Il10 in plasma, tissue extracts or cell culture supernatants.
Detection Limit 100pg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 10ng Mouse IL-10 standard, 1 vial 50X Biotinylated IL-10 Antibody, 30mL 10X MIX diluent concentrate, 30mL Wash Buffer 20X concentrate, 90μL Streptavidin-Peroxidase Conjugate (SP Conjugate),, 8mL chromog
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma, Serum, Tissue
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type Il10 (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.