Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ KDR (Human) ELISA Kit

Catalog No. p-4747566 Shop All Abnova Corporation Products
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
CCS, Plasma, Serum, Tissue Sample, Urine
Cell Lysates, Plasma, Serum
Serum, Plasma, CCS, Urine

Catalog No. 89-100-419

Supplier: Abnova™ KA2221

Only null left
Add to Cart
Add to Cart

Includes

  • 96-Well Microplate or Strips Coated w/Capture Antibody, 12 x 8 Strips
  • Ready-to-Use Streptavidin-HRP, 12mL Clear container
  • Ready-to-Use Substrate, 12mL, Brown container
  • Stop Solution, 12mL, Clear container
  • Wash Buffer (10x), 50mL, Clear container
  • Protein Standard Diluent, 12mL, Clear container
  • Sample Diluent, 12mL, Clear container
  • Detection Antibody Diluent, 12mL, Clear container
  • Biotin-Labeled Detection Antibody, Lyophilized, Yellow container
  • Protein Standard, Lyophilized (6ng), Red container
  • Adhesive Plate Sealers, 4 Sheets

This immunoassay utilizes the quantitative technique of a “Sandwich” Enzyme-Linked Immunosorbent Assay (ELISA) where the target protein (antigen) is bound in a “sandwich” format by the primary capture antibodies coated to each well-bottom and the secondary detection antibodies added subsequently by the investigator

Specifications

Conjugate Biotin
Cross Reactivity Human
Description KDR (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human KDR.
Detection Method Colorimetric
Includes Antibody coated microwells (96 wells), Biotin-conjugated detection antibody (1 vial), Avidin-HRP conjugate (10UL), Cytokine protein standard (1 vial); Ready-to-use substrate (11mL), Stop solution (11mL), Wash buffer 10X (50mL), Protein standard diluent (15mL), Detection antibody diluent (15mL), HRP diluent (15mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Lysates, Plasma, Serum
Sample Volume 100 μL
Sensitivity 32 to 2000 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.