Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Lcn2 (Mouse) ELISA Kit

Catalog No. p-4392054
Click to view available options
Sample Volume:
10 μL
100 μL

Includes

  • 96-well plate precoated with anti-mouse NGAL antibody, 96 (8 x 12) wells
  • Lyophilized recombinant mouse NGAL standard, 10 ng/tube × 2
  • Biotinylated anti- mouse NGAL antibody, dilution 1:100, 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC), dilution 1:100, 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Lcn2

Specifications

Calibration Range 78 to 5000 pg/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Lcn2 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Lcn2.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes 96-well plate precoated with Anti-Mouse NGAL Antibody, 2 x 10ng recombinant Mouse NGAL standard (lyophilized), 130μL Biotinylated Anti-Mouse NGAL Antibody, 130μL Avidin-Biotin-Peroxidase Complex (ABC), 30mL Sample Dilue
Quantity 1 Kit
Sample Type CCS, Plasma (Heparin), Serum, Urine
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type LCN2 (Mouse) ELISA kit
No. of Assays 96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.