Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ LYZ (Human) ELISA Kit

Catalog No. p-4392074 Shop All Abnova Corporation Products
Click to view available options
Sample Type:
CCS, CSF, Plasma, Saliva, Serum, Urine
Milk

Sandwich Enzyme Immunoassay for the quantitative measurement of Human LYZ

Specifications

Calibration Range 0.093 to 6μg/mL
Conjugate HRP
Cross Reactivity Human
Description LYZ (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of human LYZ.
Detection Limit 0.09μg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 6μg Human LYZ standard, 1 vial Biotinylated LYZ, 30mL 10X EIA diluent concentrate, 30mL Wash Buffer 20X concentrate, 80μL Streptavidin-Peroxidase Conjugate (SP Conjugate), 8mL chromoge
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Milk
Sample Volume 25 μL
Content And Storage Store SP Conjugate at -20°C. Store Standard and Biotinylated protein at 2-8°C before reconstituting with Diluent and at -20°C after reconstituting with Diluent. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type LYZ (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.