Learn More
Specifications
Specifications
| Content And Storage | Store the kit at -20°C. |
| Description | PGM Assay Kit (Colorimetric) is a quantitative colormetric determination of PGM at 450 nm. |
| Detection Method | Colorimetric |
| For Use With (Application) | Functional Study |
| Quantity | 1 Kit |
| Sample Type | Attached Cell, Plasma, Suspension Cell, Tissue |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.