Learn More
Description
Includes
- Phenylalanine Assay Buffer 25mL
- Tyrosinase lyophilized
- Enzyme Mix lyophilized
- Developer lyophilized
- Phenyalanine Standard (10.0μmol) lyophilized
Specifications
Specifications
| Calibration Range | 0.1 to 10nm |
| Content And Storage | Store the kit at -20°C. |
| Description | Phenylalanine Assay Kit (Fluorometric) is used for quantifying PHE. |
| Detection Method | Fluorometric |
| For Use With (Application) | Functional Study |
| Includes | 25mL Phenylalanine assay buffer, 1 vial Tyrosinase, 1 vial enzyme mix, 1 vial developer, 1 vial Phenylalanine standard |
| Quantity | 1 Kit |
| Sample Type | Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine |
| Type | Phenylalanine Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.