Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SELE (Rat) ELISA Kit

Catalog No. 89101095
Click to view available options
Quantity:
1 Kit

Includes

  • 96-well plate precoated with anti-rat E-Selectin antibody, 96 (8 x 12) wells
  • Lyophilized recombinant rat E-Selectinstandard, 10ng x 2
  • Biotinylated anti-rat E-Selectin antibody (dilution 1:100), 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100), 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

Use to measure quantitatively rat Sele. Sandwich enzyme immunoassay.

Sele (Rat) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Assay Sensitivity <10 pg/mL
Calibration Range 156 to 10,000pg/mL
Content And Storage Store the kit at -20°C
Description Sele (Rat) is a sandwich enzyme immunoassay for the quantitative measurement of rat Sele.
Conjugate Biotin
Cross Reactivity Rat
Detection Method Colorimetric
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Specificity Natural and recombinant rat sE-Selectin
Sufficient For 96 Assays
For Use With (Application) Quantification
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Detection Limit <10pg/mL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.