Learn More
Specifications
Specifications
| Content And Storage | Store the kit at 4°C temporarily. For best results, store the components as described in the protocol. |
| Description | Steatosis Colorimetric Assay Kit provides a method for evaluating steatosis risk of drug candidates using Oil Red O to stain neutral lipids in hepatocytes. |
| Detection Method | Colorimetric |
| For Use With (Application) | Functional Study |
| Includes | 10mL Fixative 10X, 30mL wash solution, 25mL Oil Red O solution, 30mL dye extraction solution, 100μL Chloroquine positive control (25mM), 30mL Steatosis assay Hematoxylin |
| Quantity | 1 Kit |
| Type | Detection Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.