Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Treponema pallidum IgG ELISA Kit
Click to view available options
Quantity:
96 Tests
Specifications
Specifications
Conjugate | HRP |
Description | Treponema pallidum IgG ELISA Kit is used for the detection of IgG antibody to Treponema pallidum in human serum or plasma. |
Detection Method | Colorimetric |
Includes | Microwell 12 x 8 x 1, 22mL sample diluent, 1.5mL calibrator, 1.5mL positive control, 1.5mL negative control, 12mL Enzyme Conjugate, 12mL TMB chromogenic Substrate, 12mL stop solution, 25mL Washing concentrate (20X) |
Quantity | 96 Tests |
Regulatory Status | For research use only (RUO) |
Sample Type | Plasma, Serum |
Sample Volume | 10 μL |
Content And Storage | Store the kit at 4°C |
For Use With (Application) | Qualitative |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction